Anti-BSA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: US Biological
A1275-02A
USBIA1275-02AEA 11275 NOK
USBIA1275-02A
Anti-BSA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: Western Blot: 1:2000-1:10000; ELISA: 1:20000-1:80000; Immunochemistry: 1:500-1:2500

Type: Primary
Antigen: BSA (Bovine Serum Albumin)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR