Anti-BSA Rabbit Polyclonal Antibody

Leverandør: US Biological
A1275-03B
USBIA1275-03BEA 6900 NOK
USBIA1275-03B
Anti-BSA Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0. 02-0.1 ?g/ml

Type: Primary
Antigen: BSA (Bovine Serum Albumin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol BSA
Antigen-navn Bovine Serum Albumin
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Isotype IgG
Western blot Yes
Skjema liquid
Rensing purified by Protein A chromatography
Konsentrasjon ~0,5 mg/ml
Lagringsbuffer PBS, 0,05% BSA, 0,05% sodium azide.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR