Anti-BSA Chicken Polyclonal Antibody

Leverandør: US Biological
A1275-04A
USBIA1275-04AEA 5425 NOK
USBIA1275-04A
Anti-BSA Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:10000

Type: Primary
Antigen: BSA (Bovine Serum Albumin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Bovine
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR