Anti-BSA Rabbit Polyclonal Antibody

Leverandør: US Biological
A1275-04H
USBIA1275-04HEA 6725 NOK
USBIA1275-04H
Anti-BSA Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:1000-1:5000; ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000

Type: Primary
Antigen: BSA (Bovine Serum Albumin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR