Anti-BSA Mouse Monoclonal Antibody [clone: 6F10]

Leverandør: US Biological
A1275-06C
USBIA1275-06CEA 9850 NOK
USBIA1275-06C
Anti-BSA Mouse Monoclonal Antibody [clone: 6F10]
Antistoffer
Recommended dilution: ELISA: 1:1000

Type: Primary
Antigen: BSA (Bovine Serum Albumin)
Clonality: monoclonal
Clone: 6F10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR