Anti-BSA Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
A1325-20B
USBIA1325-20BEA 5975 NOK
USBIA1325-20B
Anti-BSA Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 1:2000-1:10000; Immunochemistry: 1:100-1:250;

Type: Primary
Antigen: BSA (Bovine Serum Albumin)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Goat, Guinea pig, Horse, Human, Monkey, Pig, Rat, Sheep
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR