Anti-ACE/AGT Rabbit Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
A2294-48V
USBIA2294-48VEA 14275 NOK
USBIA2294-48V
Anti-ACE/AGT Rabbit Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: ACE/AGT (Angiotensin I, II)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR