Anti-ART1, ID GPI-linked NAD Rabbit Polyclonal Antibody

Leverandør: US Biological
A3577-90A
USBIA3577-90AEA 8200 NOK
USBIA3577-90A
Anti-ART1, ID GPI-linked NAD Rabbit Polyclonal Antibody
Antistoffer
  • Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: ART1, ID GPI-linked NAD
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol ART1, ID GPI-linked NAD
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Skjema liquid
Rensing purified by Protein G chromatography
Lagringsbuffer PBS, 0,09% sodium azide.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR