Anti-ART1, ID GPI-linked NAD Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: US Biological
A3577-90A-AP
USBIA3577-90A-APEA 10875 NOK
USBIA3577-90A-AP
Anti-ART1, ID GPI-linked NAD Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: ART1, ID GPI-linked NAD
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol ART1, ID GPI-linked NAD
Antistofftype Primary
Klonalitet Polyclonal
Konjugering AP (Alkaline Phosphatase)
Reaktivitet Human
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Skjema liquid
Rensing purified by Protein G chromatography
Lagringsbuffer PBS, pH 7,2, contains ~0,09% sodium azide
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR