Anti-ART1, ID GPI-linked NAD Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: US Biological
A3577-90A-APC
USBIA3577-90A-APCEA 10875 NOK
USBIA3577-90A-APC
Anti-ART1, ID GPI-linked NAD Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: ART1, ID GPI-linked NAD
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol ART1, ID GPI-linked NAD
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering APC (Allophycocyanin)
Reaktivitet Human
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Skjema liquid
Rensing purified by Protein G chromatography
Lagringsbuffer PBS, pH 7,2, contains ~0,09% sodium azide
Lagringstemp Cool
Frakttemp. Ice
Uttestet applikasjon Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR