Anti-ART1, ID GPI-linked NAD Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
A3577-90A-BIOT
USBIA3577-90A-BIOTEA 10875 NOK
USBIA3577-90A-BIOT
Anti-ART1, ID GPI-linked NAD Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: ART1, ID GPI-linked NAD
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR