Anti-ART1, ID GPI-linked NAD Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: US Biological
A3577-90A-FITC
USBIA3577-90A-FITCEA 10875 NOK
USBIA3577-90A-FITC
Anti-ART1, ID GPI-linked NAD Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: ART1, ID GPI-linked NAD
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR