Anti-Bcl 10, Mouse Monoclonal Antibody (KLH (Keyhole Limpet Hemocyanin)) [clone: 6D158]

Leverandør: US Biological
B0807-12F
USBIB0807-12FEA 9625 NOK
USBIB0807-12F
Anti-Bcl 10, Mouse Monoclonal Antibody (KLH (Keyhole Limpet Hemocyanin)) [clone: 6D158]
Antistoffer
Recommended dilution: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: Bcl 10,
Clonality: Monoclonal
Clone: 6D158
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,
Bestill nå

SPESIFIKASJONER

Antigensymbol BCL10
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering KLH (Keyhole Limpet Hemocyanin)
Klon 6D158
Reaktivitet Human
Vert Mouse
Isotype IgG1
Western blot Yes
Modifikasjon phospho-Ser138
Immunogen Synthetic peptide corresponding to a portion of Bcl10 phosphorylated at Ser138
Skjema liquid
Rensing purified by thiophilic adsorption and size exclusion chromatography
Kyssabsorpsjon No
Lagringsbuffer PBS, 0,09% sodium azide, PEG, Sucrose and 40% glycerol.
Lagringstemp –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR