Anti-Bilirubin Oxidase Goat Polyclonal Antibody

Leverandør: US Biological
B1140-06
USBIB1140-06EA 11575 NOK
USBIB1140-06
Anti-Bilirubin Oxidase Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:2000 -1:10000; Western Blot: 1:500-1:5000; Immunoprecipitation: 1:100

Type: Primary
Antigen: Bilirubin Oxidase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR