Anti-Biotin Goat Polyclonal Antibody (TRITC (Rhodamine))

Leverandør: US Biological
B1750-01P
USBIB1750-01PEA 3650 NOK
USBIB1750-01P
Anti-Biotin Goat Polyclonal Antibody (TRITC (Rhodamine))
Antistoffer
Recommended dilution: Flow Cytometry: 1:20-1:2000

Type: Primary
Antigen: Biotin (biotin)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR