Anti-Biotin Goat Polyclonal Antibody

Leverandør: US Biological
B1750-28A
USBIB1750-28AEA 6475 NOK
USBIB1750-28A
Anti-Biotin Goat Polyclonal Antibody
Antistoffer
Type: Tag
Antigen: Biotin (biotin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR