Anti-Biotin Goat Polyclonal Antibody (TRITC (Rhodamine))

Leverandør: US Biological
B1750-37
USBIB1750-37EA 15000 NOK
USBIB1750-37
Anti-Biotin Goat Polyclonal Antibody (TRITC (Rhodamine))
Antistoffer
Type: Primary
Antigen: Biotin (biotin)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype:
Reactivity: Goat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR