Anti-Catestatin Rabbit Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
C2096-86E6
USBIC2096-86E6EA 12100 NOK
USBIC2096-86E6
Anti-Catestatin Rabbit Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: Catestatin
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR