Anti-CD3E Rabbit Polyclonal Antibody

Leverandør: US Biological
C2254-18C
USBIC2254-18CEA 8300 NOK
USBIC2254-18C
Anti-CD3E Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: CD3E (CD3e molecule, epsilon (CD3-TCR complex))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD3E
Environmentally Preferable
Antigen-navn CD3e molecule, epsilon (CD3-TCR complex)
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Isotype
Aminosyretall 156 to 168
Skjema liquid
Rensing purified by immunoaffinity chromatography
Lagringsbuffer 10mM PBS, 1% BSA, pH 7,4, 0,1% sodium azide.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR