Anti-CD14 Mouse Monoclonal Antibody [clone: 11E72]

Leverandør: US Biological
C2265-55BX
USBIC2265-55BXEA 8375 NOK
USBIC2265-55BX
Anti-CD14 Mouse Monoclonal Antibody [clone: 11E72]
Antistoffer
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: Monoclonal
Clone: 1.1E+73
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD14
Environmentally Preferable
Antigen-navn CD14 molecule
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon 11E72
Reaktivitet Human
Vert Mouse
Isotype IgG1 kappa
Flowcytometri Yes
Skjema liquid
Rensing purified by Protein G chromatography
Konsentrasjon ~0,5 mg/ml
Lagringsbuffer PBS
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR