Anti-CD14 Rat Monoclonal Antibody [clone: Sa14-2]

Leverandør: US Biological
C2265-55C
USBIC2265-55CEA 12550 NOK
USBIC2265-55C
Anti-CD14 Rat Monoclonal Antibody [clone: Sa14-2]
Antistoffer
Recommended dilution: Flow Cytometry: 1 ?g/ml; Immunofluorescence/; Immunochemistry: 2?g/ml

Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: Monoclonal
Clone: Sa14-2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR