Anti-Choline Acetyltransferase Rabbit Polyclonal Antibody

Leverandør: US Biological
C5057-02E
USBIC5057-02EEA 7400 NOK
USBIC5057-02E
Anti-Choline Acetyltransferase Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 1:200

Type: Primary
Antigen: Choline Acetyltransferase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol ChAT
Antigen-navn Choline Acetyltransferase
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype
Western blot Yes
Skjema liquid
Rensing purified by immunoaffinity chromatography
Konsentrasjon ~0,1 mg/ml
Lagringsbuffer PBS, 0,1M Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR