Anti-Choline Acetyltransferase Chicken Polyclonal Antibody

Leverandør: US Biological
C5057-06B
USBIC5057-06BEA 3975 NOK
USBIC5057-06B
Anti-Choline Acetyltransferase Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:250 -1:500; Immunochemistry: 1:100 -1:200

Type: Primary
Antigen: Choline Acetyltransferase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR