Anti-Colon Carcinoma Mouse Monoclonal Antibody [clone: CC-1]

Leverandør: US Biological
C7554-50
USBIC7554-50EA 6525 NOK
USBIC7554-50
Anti-Colon Carcinoma Mouse Monoclonal Antibody [clone: CC-1]
Antistoffer
Recommended dilution: Immunochemistry: 1:10 Stains acetone or ethanol fixed tissues

Type: Primary
Antigen: Colon Carcinoma
Clonality: Monoclonal
Clone: CC-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR