Anti-Complement C3c Pig Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
C7850-14C6
USBIC7850-14C6EA 6325 NOK
USBIC7850-14C6
Anti-Complement C3c Pig Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: Complement C3c
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Pig
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR