Anti-CRK Mouse polyclonal antibody

Leverandør: US Biological
C7924-10J
USBIC7924-10JEA 8050 NOK
USBIC7924-10J
Anti-CRK Mouse polyclonal antibody
Antistoffer
Type: Primary
Antigen: CRK (v-crk avian sarcoma virus CT10 oncogene homolog)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR