Anti-DHEA Mouse monoclonal antibody [clone: 1­25.1 (BGN/1177/1251)]

Leverandør: US Biological
D3224-98F
USBID3224-98FEA 7250 NOK
USBID3224-98F
Anti-DHEA Mouse monoclonal antibody [clone: 1­25.1 (BGN/1177/1251)]
Antistoffer
  • Recommended dilution: ELISA: 1:10000

Type: Primary
Antigen: DHEA
Clonality: monoclonal
Clone: 1­25.1 (BGN/1177/1251)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR