Drop-Out Mix Complete without Yeast Nitrogen Base
Leverandør: US Biological
D9515-100G
USBID9515-100GEA 2500 NOK
USBID9515-100G
Drop-Out Mix Complete without Yeast Nitrogen Base
Mikrobiologiske medier Mikrobiologiske tilsetninger og supplementer
Drop-out mixes are used as a supplement to Drop-out Base for making synthetic, complete media formulations in accordance with methods in yeast genetics.
  • Convenient powders
  • Useful for one-hybrid and two-hybrid systems
All lots are quality control tested using the appropriate yeast strains and transformants.