Drop-out mix synthetic minus adenine, histidine, leucine, tryptophan, uracil without yeast nitrogen base
Leverandør: US Biological
D9540-05
USBID9540-05EA 2500 NOK
USBID9540-05
Drop-out mix synthetic minus adenine, histidine, leucine, tryptophan, uracil without yeast nitrogen base
Mikrobiologiske medier Mikrobiologiske tilsetninger og supplementer
Drop-out mixes are used as a supplement to Drop-out base for making synthetic, complete media formulations in accordance with methods in yeast genetics.
  • Convenient powders
  • Useful for one-hybrid and two-hybrid systems
All lots are quality control tested using the appropriate yeast strains and transformants.