Anti-Estradiol, 17 beta Mouse Monoclonal Antibody [clone: 6F92]

Leverandør: US Biological
E3550-20
USBIE3550-20EA 10475 NOK
USBIE3550-20
Anti-Estradiol, 17 beta Mouse Monoclonal Antibody [clone: 6F92]
Antistoffer
Recommended dilution: ELISA: 1:4000

Type: Primary
Antigen: Estradiol, 17 beta
Clonality: monoclonal
Clone: 6F92
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR