Anti-Exendin 4 Rabbit polyclonal antibody Rhodamine

Leverandør: US Biological
E9050-03E4
USBIE9050-03E4EA 14275 NOK
USBIE9050-03E4
Anti-Exendin 4 Rabbit polyclonal antibody Rhodamine
Antistoffer
Type: Primary
Antigen: Exendin 4
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR