Anti-Fluorescein Isothiocyanate Sheep Polyclonal Antibody (Biotin)

Leverandør: US Biological
F5250-11J2
USBIF5250-11J2EA 7875 NOK
USBIF5250-11J2
Anti-Fluorescein Isothiocyanate Sheep Polyclonal Antibody (Biotin)
Antistoffer
Type: Tag
Antigen: Fluorescein Isothiocyanate
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol FITC
Environmentally Preferable
Antigen-navn Fluorescein isothiocyanate
Antistofftype Tag
Klonalitet Polyclonal
Konjugering Biotin
Reaktivitet
Vert Sheep
Isotype IgG
ELISA Yes
Skjema liquid
Rensing purified by affinity chromatography
Konsentrasjon ~0,5 mg/ml
Lagringsbuffer PBS, 0,09% sodium azide
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR