Anti-FOXD4L1 Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))

Leverandør: US Biological
F9046-17-100 F9046-17-50
USBIF9046-17-100EA 3325 NOK
USBIF9046-17-100 USBIF9046-17-50
Anti-FOXD4L1 Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))
Antistoffer
Recommended dilution: ELISA: 1: 250000

Type: Primary
Antigen: FOXD4L1
Clonality: Polyclonal
Clone:
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Bestill nå

SPESIFIKASJONER

Antigensymbol FOXD4L1 FOXD4L1
Environmentally Preferable
Antistofftype Primary Primary
Klonalitet Polyclonal Polyclonal
Konjugering KLH (Keyhole Limpet Hemocyanin) KLH (Keyhole Limpet Hemocyanin)
Reaktivitet Human Human
Vert Rabbit Rabbit
Isotype IgG IgG
ELISA Yes Yes
Immunogen Synthetic peptide (C-GLRQRLRSHQGRGAG) corresponding to human FOXD4L1 (KLH). Molecular Weight: 43.6kD. Synthetic peptide (C-GLRQRLRSHQGRGAG) corresponding to human FOXD4L1 (KLH). Molecular Weight: 43.6kD.
Sekvens C-GLRQRLRSHQGRGAG
Skjema liquid liquid
Rensing purified by immunoaffinity chromatography purified by immunoaffinity chromatography
Konsentrasjon ~0,5 mg/ml ~0,5 mg/ml
Kyssabsorpsjon No No
Lagringsbuffer PBS, 0,02% sodium azide, before the addition of glycerol to 40%. PBS, 0,02% sodium azide, before the addition of glycerol to 40%.
Lagringstemp –20 °C –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR