Anti-FOXE1 Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))

Leverandør: US Biological
F9046-25A-100
USBIF9046-25A-100EA 3325 NOK
USBIF9046-25A-100
Anti-FOXE1 Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))
Antistoffer
Recommended dilution: ELISA: 1: 250000

Type: Primary
Antigen: FOXE1
Clonality: Polyclonal
Clone:
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Bestill nå

SPESIFIKASJONER

Antigensymbol FOXE1
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering KLH (Keyhole Limpet Hemocyanin)
Reaktivitet Human
Vert Rabbit
Isotype IgG
ELISA Yes
Immunogen Synthetic peptide (TGARPANPSAYAAAY-C) corresponding to human FOXE1 (KLH). Molecular Weight: 38.2kD.
Sekvens TGARPANPSAYAAAYC
Skjema liquid
Rensing purified by immunoaffinity chromatography
Konsentrasjon ~0,25 mg/ml
Kyssabsorpsjon No
Lagringsbuffer PBS, 0,02% sodium azide, before the addition of glycerol to 40%.
Lagringstemp –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR