Anti-FOXO4 Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))

Leverandør: US Biological
F9049-45-100
USBIF9049-45-100EA 3325 NOK
USBIF9049-45-100
Anti-FOXO4 Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))
Antistoffer
Recommended dilution: ELISA: 1: 75000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: FOXO4
Clonality: Polyclonal
Clone:
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR