Anti-GHRL Rabbit Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
G2033-74T3
USBIG2033-74T3EA 14275 NOK
USBIG2033-74T3
Anti-GHRL Rabbit Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: GHRL (ghrelin/obestatin prepropeptide)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR