Anti-Glyoxalase Rabbit polyclonal antibody

Leverandør: US Biological
G8235-29W
USBIG8235-29WEA 7275 NOK
USBIG8235-29W
Anti-Glyoxalase Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: Glyoxalase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn Glyoxalase
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Skjema lyophilized powder
Rensing purified by solid phase affinity chromatography
Lagringsbuffer PBS, pH 7,2, Dextran.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR