Anti-Glyoxylate Reductase Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
G8235-36F
USBIG8235-36FEA 6175 NOK
USBIG8235-36F
Anti-Glyoxylate Reductase Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Glyoxylate Reductase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol GLYR
Antigen-navn Glyoxylate Reductase
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Biotin
Reaktivitet
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Skjema lyophilized powder
Rensing purified by ammonium sulphate precipitation and ion exchange chromatography
Lagringsbuffer PBS, pH 7,2. No
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR