Anti-G Protein Coupled Receptor 171 Rabbit polyclonal antibody

Leverandør: US Biological
G8601-75
USBIG8601-75EA 8375 NOK
USBIG8601-75
Anti-G Protein Coupled Receptor 171 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: G Protein Coupled Receptor 171
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR