Anti-GFP Goat Polyclonal Antibody (TRITC (Rhodamine))

Leverandør: US Biological
G8965-13
USBIG8965-13EA 15375 NOK
USBIG8965-13
Anti-GFP Goat Polyclonal Antibody (TRITC (Rhodamine))
Antistoffer
Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR