Anti-IgG, Fc Goat Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
I1903-19N5
USBII1903-19N5EA 4400 NOK
USBII1903-19N5
Anti-IgG, Fc Goat Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: IgG, Fc
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR