Anti-IgG Goat Polyclonal Antibody (FAM (Carboxyfluorescein))

Leverandør: US Biological
I1903-56B
USBII1903-56BEA 5825 NOK
USBII1903-56B
Anti-IgG Goat Polyclonal Antibody (FAM (Carboxyfluorescein))
Antistoffer
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering FAM (Carboxyfluorescein)
Reaktivitet Mouse
Vert Goat
Isotype IgG
ImmunoFluorescens Yes
Flowcytometri Yes
Skjema liquid
Rensing purified by affinity chromatography
Lagringsbuffer PBS, 0,5% BSA, 0,1% sodium azide.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR