Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
I1903-62W
USBII1903-62WEA 5050 NOK
USBII1903-62W
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Recommended dilution: Flow Cytometry: 1:20-1:2000

Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR