Anti-IgG Rabbit Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
I1903-88A7
USBII1903-88A7EA 5000 NOK
USBII1903-88A7
Anti-IgG Rabbit Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR