Anti-IgG Goat Polyclonal Antibody (TRITC (Rhodamine))

Leverandør: US Biological
I1903-89U7
USBII1903-89U7EA 2900 NOK
USBII1903-89U7
Anti-IgG Goat Polyclonal Antibody (TRITC (Rhodamine))
Antistoffer
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR