Anti-IgG, F Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
I1904-41Z
USBII1904-41ZEA 6400 NOK
USBII1904-41Z
Anti-IgG, F Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgG, F
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR