Anti-IgG Chicken Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
I1904-45L7
USBII1904-45L7EA 3250 NOK
USBII1904-45L7
Anti-IgG Chicken Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR