Anti-IgG/IgM Goat Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
I1904-48G7
USBII1904-48G7EA 3175 NOK
USBII1904-48G7
Anti-IgG/IgM Goat Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: IgG/IgM (IgG/IgM)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR