Anti-IgG Goat Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
I1904-53E6
USBII1904-53E6EA 3950 NOK
USBII1904-53E6
Anti-IgG Goat Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR