Anti-IgM Goat Polyclonal Antibody (TRITC (Rhodamine))

Leverandør: US Biological
I1905-04C7
USBII1905-04C7EA 2550 NOK
USBII1905-04C7
Anti-IgM Goat Polyclonal Antibody (TRITC (Rhodamine))
Antistoffer
Type: Primary
Antigen: IgM (IgM)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR